Gene Transcripts

Gene Symbol
Rhoh
Entrez Gene ID
305341
Gene Name
ras homolog family member H
Gene Aliases
Chromosome Location
Chr. 14: 43961756 - 43992587 on Build Rnor_6.0
UniGene ID
Rn.20618
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013430.1 NP_001013448.1 3 795
GenBank mRNA BC091269.1 - 3 795
FQ225173.1 - 3 807
FQ232983.1 - 3 806

View Full Product Details