Gene Transcripts

Gene Symbol
St8sia5
Entrez Gene ID
364901
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5
Gene Aliases
SIAT8E
Chromosome Location
Chr. 18: 73564034 - 73629016 on Build Rnor_6.0
UniGene ID
Rn.199050
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213628.1 NP_998793.1 6 595
XM_006254968.2 XP_006255030.1 7 981
GenBank mRNA AJ699422.1 - 6 595

View Full Product Details