Gene Transcripts

Gene Symbol
Serpinb2
Entrez Gene ID
60325
Gene Name
serpin peptidase inhibitor, clade B (ovalbumin), member 2
Gene Aliases
Pai2a
Chromosome Location
Chr. 13: 27449895 - 27463015 on Build Rnor_6.0
UniGene ID
Rn.42912
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021696.2 NP_067728.1 4 493
XM_006249639.2 XP_006249701.1 5 679
XM_008769495.1 XP_008767717.1 5 576
GenBank mRNA X64563.1 - 4 494

View Full Product Details