Gene Transcripts

Gene Symbol
Ctbs
Entrez Gene ID
81652
Gene Name
chitobiase, di-N-acetyl-
Gene Aliases
Chromosome Location
Chr. 2: 252305874 - 252320387 on Build Rnor_6.0
UniGene ID
Rn.11199
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031023.1 NP_112285.1 3 428
GenBank mRNA BC088129.1 - 3 431
FQ219359.1 - 3 437
FQ229296.1 - 1 51
M95768.1 - 3 428

View Full Product Details