Gene Transcripts

Gene Symbol
Gsto1
Entrez Gene ID
114846
Gene Name
glutathione S-transferase omega 1
Gene Aliases
SPG-R
Chromosome Location
Chr. 1: 267607437 - 267617387 on Build Rnor_6.0
UniGene ID
Rn.25166
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007602.1 NP_001007603.1 6 721
GenBank mRNA BC079363.1 - 6 721
FQ218105.1 - 6 696
FQ220742.1 - 6 663

View Full Product Details