Gene Transcripts

Gene Symbol
Crtc3
Entrez Gene ID
365297
Gene Name
CREB regulated transcription coactivator 3
Gene Aliases
RGD1309666
Chromosome Location
Chr. 1: 142398248 - 142500558 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229462.2 XP_006229524.1 15 1968
XM_006229463.2 XP_006229525.1 15 1965

Gene Symbol
LOC100909455
Entrez Gene ID
100909455
Gene Name
CREB-regulated transcription coactivator 3-like
Gene Aliases
Chromosome Location
Chr. 1: 142388430 - 142396313 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748889.3 XP_003748937.2 1631
XM_006229461.2 XP_006229523.1 1629

View Full Product Details