Gene Transcripts

Gene Symbol
Shd
Entrez Gene ID
316507
Gene Name
src homology 2 domain-containing transforming protein D
Gene Aliases
Chromosome Location
Chr. 9: 11090048 - 11098826 on Build Rnor_6.0
UniGene ID
Rn.11845
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108223.1 NP_001101693.1 3 578
XM_006244329.1 XP_006244391.1 3 2667
GenBank mRNA BC158646.1 - 3 520

View Full Product Details