Gene Transcripts

Gene Symbol
Abtb1
Entrez Gene ID
297432
Gene Name
ankyrin repeat and BTB (POZ) domain containing 1
Gene Aliases
Chromosome Location
Chr. 4: 120778580 - 120784807 on Build Rnor_6.0
UniGene ID
Rn.40721
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005902.1 NP_001005902.1 10 1056
GenBank mRNA BC083579.1 - 10 1056

View Full Product Details