Gene Transcripts

Gene Symbol
Clpp
Entrez Gene ID
301117
Gene Name
caseinolytic mitochondrial matrix peptidase proteolytic subunit
Gene Aliases
Chromosome Location
Chr. 9: 10048715 - 10056075 on Build Rnor_6.0
UniGene ID
Rn.12681
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_217313.8 XP_217313.6 4 2106
GenBank mRNA FQ231323.1 - 4 393

View Full Product Details