Gene Transcripts

Gene Symbol
Lnx2
Entrez Gene ID
360761
Gene Name
ligand of numb-protein X 2
Gene Aliases
Chromosome Location
Chr. 12: 9728378 - 9793428 on Build Rnor_6.0
UniGene ID
Rn.162084
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108329.1 NP_001101799.1 2 464
XM_006248835.2 XP_006248897.1 2 591
XM_008768976.1 XP_008767198.1 1 5510

View Full Product Details