Gene Transcripts

Gene Symbol
Col13a1
Entrez Gene ID
499431
Gene Name
collagen, type XIII, alpha 1
Gene Aliases
Chromosome Location
Chr. 20: 31456502 - 31598356 on Build Rnor_6.0
UniGene ID
Rn.17566
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109172.1 NP_001102642.1 14 1179
XM_006256470.2 XP_006256532.2 14 1236

View Full Product Details