Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001572.3 | NP_001563.2 |
NM_004029.2 | NP_004020.1 | |
NM_004031.2 | NP_004022.2 | |
XM_005252906.3 | XP_005252963.1 | |
XM_005252907.3 | XP_005252964.1 | |
XM_005252909.3 | XP_005252966.1 | |
XM_011520066.2 | XP_011518368.1 | |
XM_017017674.1 | XP_016873163.1 | |
GenBank mRNA | AF076494.1 | - |
AK303752.1 | - | |
BC136555.1 | - | |
BC144242.1 | - | |
BM849172.1 | - | |
DA601567.1 | - | |
DA967682.1 | - | |
ENST00000330243.0 | - | |
KC477210.1 | - | |
U53830.1 | - | |
U53831.1 | - | |
U53832.1 | - | |
U73036.1 | - |