Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001252600.1 | NP_001239529.1 |
NM_001252601.1 | NP_001239530.1 | |
NM_016850.3 | NP_058546.1 | |
GenBank mRNA | AK002830.1 | - |
AK004349.1 | - | |
AK079685.1 | - | |
AK089869.1 | - | |
AK150403.1 | - | |
AK150608.1 | - | |
AK151470.1 | - | |
AK152401.1 | - | |
AK152403.1 | - | |
AK153054.1 | - | |
AK155950.1 | - | |
AK158378.1 | - | |
AK158403.1 | - | |
AK159142.1 | - | |
AK159883.1 | - | |
BC138799.1 | - | |
BC145422.1 | - | |
BQ033099.1 | - | |
BY155234.1 | - | |
DT923384.1 | - | |
U73037.1 | - |