Gene Transcripts

Gene Symbol
Irf7
Entrez Gene ID
54123
Gene Name
interferon regulatory factor 7
Gene Aliases
-
Chromosome Location
Chr.7: 141263183 - 141266499 on Build GRCm38
Uni Gene
Mm.3233
Interrogated Sequence Translated Protein
RefSeq NM_001252600.1 NP_001239529.1
NM_001252601.1 NP_001239530.1
NM_016850.3 NP_058546.1
GenBank mRNA AK002830.1 -
AK004349.1 -
AK079685.1 -
AK089869.1 -
AK150403.1 -
AK150608.1 -
AK151470.1 -
AK152401.1 -
AK152403.1 -
AK153054.1 -
AK155950.1 -
AK158378.1 -
AK158403.1 -
AK159142.1 -
AK159883.1 -
BC138799.1 -
BC145422.1 -
BQ033099.1 -
BY155234.1 -
DT923384.1 -
U73037.1 -

View Full Product Details