Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001278601.1 | NP_001265530.1 |
NM_013693.3 | NP_038721.1 | |
GenBank mRNA | AK153319.1 | - |
AK153800.1 | - | |
AK154223.1 | - | |
AK155964.1 | - | |
AY423855.1 | - | |
BC117057.1 | - | |
BC137720.1 | - | |
BY167004.1 | - | |
M11731.1 | - | |
M13049.1 | - | |
X02611.1 | - |