Gene Transcripts
- Gene Symbol
- 4632415L05Rik
- Entrez Gene ID
- 70808
- Gene Name
- RRS1 ribosome biogenesis regulator homolog pseudogene
- Gene Aliases
- AI662792
- Chromosome Location
- Chr. 3: 19894871 - 19898984 on Build GRCm38
- UniGene ID
- Mm.427103
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027985.1 |
|
1 |
1697 |
|
GenBank mRNA |
AK014580.1 |
- |
1 |
1660 |
|
BC037267.1 |
- |
1 |
1670 |
|
View Full Product Details