Gene Transcripts
- Gene Symbol
- HIST2H2BA
- Entrez Gene ID
- 337875
- Gene Name
- histone cluster 2, H2ba (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 1: 121108441 - 121117251 on Build GRCh38
- UniGene ID
- Hs.742189
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027337.1 |
|
2 |
408 |
|
GenBank mRNA |
BC106916.2 |
- |
2 |
408 |
|
BC126904.1 |
- |
2 |
409 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CF887452.1 |
- |
2 |
414 |
|
Chr. 1 - 121108210 - 121117257 [-] |
View Full Product Details