Gene Transcripts
- Gene Symbol
- LINC01102
- Entrez Gene ID
- 150568
- Gene Name
- long intergenic non-protein coding RNA 1102
- Gene Aliases
- Chromosome Location
- Chr. 2: 104434347 - 104512757 on Build GRCh38
- UniGene ID
- Hs.107284
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX460258.2 |
- |
3 |
605 |
|
Chr. 2 - 104434482 - 104512361 [+] |
GenBankmRNA |
BM925825.1 |
- |
3 |
727 |
|
Chr. 2 - 104434381 - 104512303 [+] |
View Full Product Details