Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_027113.2 | 2, 3 | 614 | ||
NR_027114.2 | 2, 3 | 1019 | |||
NR_110149.1 | 2, 3 | 356 | |||
GenBank mRNA | AK093151.1 | - | 2, 3 | 987 | |
AK098520.1 | - | 2, 3 | 614 | ||
BC033369.1 | - | 2, 3 | 1019 | ||
HY000603.1 | - | 2, 3 | 325 | ||
HY207533.1 | - | 2, 3 | 287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU570925.1 | - | 327 | Chr. - - [-] |