Gene Transcripts

Gene Symbol
ROPN1L
Entrez Gene ID
83853
Gene Name
rhophilin associated tail protein 1 like
Gene Aliases
ASP, RSPH11
Chromosome Location
Chr. 5: 10441862 - 10482807 on Build GRCh38
UniGene ID
Hs.381089
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK026656.1 - 1 1121

Gene Symbol
LOC105369214
Entrez Gene ID
105369214
Gene Name
uncharacterized LOC105369214
Gene Aliases
Chromosome Location
Chr. 5: 10462865 - 10464841 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011514190.2 XP_011512492.1 1 1427

View Full Product Details