Gene Transcripts
- Gene Symbol
- SLC14A2-AS1
- Entrez Gene ID
- 101927980
- Gene Name
- SLC14A2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 18: 45438183 - 45507036 on Build GRCh38
- UniGene ID
- Hs.591058
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110899.1 |
|
7 |
2160 |
|
GenBank mRNA |
AK126075.1 |
- |
7 |
2160 |
|
View Full Product Details