Gene Transcripts
- Gene Symbol
- LINC01187
- Entrez Gene ID
- 100507267
- Gene Name
- long intergenic non-protein coding RNA 1187
- Gene Aliases
- CTB-27N1.1, KIDR
- Chromosome Location
- Chr. 5: 170191647 - 170199141 on Build GRCh38
- UniGene ID
- Hs.634846
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_108022.1 |
|
5 |
1275 |
|
GenBank mRNA |
AK055811.1 |
- |
4 |
2186 |
|
View Full Product Details