Gene Transcripts
- Gene Symbol
- LOC284244
- Entrez Gene ID
- 284244
- Gene Name
- uncharacterized LOC284244
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.4267
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF070541.1 |
- |
1 |
1168 |
|
- Gene Symbol
- ASXL3
- Entrez Gene ID
- 80816
- Gene Name
- additional sex combs like 3, transcriptional regulator
- Gene Aliases
- BRPS, KIAA1713
- Chromosome Location
- Chr. 18: 33578577 - 33751195 on Build GRCh38
- UniGene ID
- Hs.464876
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA989222.1 |
- |
1 |
255 |
|
Chr. 18 - 33750549 - 33750932 [+] |
GenBankmRNA |
BF446799.1 |
- |
1 |
221 |
|
Chr. 18 - 33750583 - 33751123 [+] |
GenBankmRNA |
BF110966.1 |
- |
1 |
256 |
|
Chr. 18 - 33750549 - 33751118 [+] |
View Full Product Details