Gene Transcripts
- Gene Symbol
- LINC01254
- Entrez Gene ID
- 101927350
- Gene Name
- long intergenic non-protein coding RNA 1254
- Gene Aliases
- Chromosome Location
- Chr. 18: 10405133 - 10414370 on Build GRCh38
- UniGene ID
- Hs.572523
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110775.1 |
|
1 |
944 |
|
GenBank mRNA |
AY726567.1 |
- |
1 |
944 |
|
BC040323.1 |
- |
1 |
1089 |
|
View Full Product Details