Gene Transcripts
- Gene Symbol
- Gm17473
- Entrez Gene ID
- 100503767
- Gene Name
- predicted gene, 17473
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.385044
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK036506.1 |
- |
2 |
837 |
|
AK138307.1 |
- |
2 |
818 |
|
View Full Product Details