Gene Transcripts
- Gene Symbol
- Gm31126
- Entrez Gene ID
- 102633258
- Gene Name
- predicted gene, 31126
- Gene Aliases
- Chromosome Location
- Chr. 13: 50267847 - 50289832 on Build GRCm38
- UniGene ID
- Mm.50673
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK030049.1 |
- |
|
488 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB616505.1 |
- |
4 |
488 |
|
Chr. 13 - 50267976 - 50392014 [+] |
View Full Product Details