Gene Transcripts
- Gene Symbol
- Adcy9
- Entrez Gene ID
- 11515
- Gene Name
- adenylate cyclase 9
- Gene Aliases
- ACtp10, AW125421, D16Wsu65e, mKIAA0520
- Chromosome Location
- Chr. 16: 4284886 - 4437236 on Build GRCm38
- UniGene ID
- Mm.439750
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK134627.1 |
- |
1 |
285 |
|
View Full Product Details