Gene Transcripts
- Gene Symbol
- 4921524L21Rik
- Entrez Gene ID
- 70901
- Gene Name
- RIKEN cDNA 4921524L21 gene
- Gene Aliases
- Chromosome Location
- Chr. 18: 6603633 - 6640435 on Build GRCm38
- UniGene ID
- Mm.158484
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK161257.1 |
- |
|
2542 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV257521.2 |
- |
1 |
291 |
|
Chr. 18 - 6640370 - 6641041 [+] |
View Full Product Details