Gene Transcripts
- Gene Symbol
- E130102H24Rik
- Entrez Gene ID
- 77866
- Gene Name
- RIKEN cDNA E130102H24 gene
- Gene Aliases
- Chromosome Location
- Chr. 4: 101346524 - 101356248 on Build GRCm38
- UniGene ID
- Mm.137053
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040708.1 |
|
2 |
581 |
|
GenBank mRNA |
AK021368.1 |
- |
2 |
543 |
|
View Full Product Details