Gene Transcripts
- Gene Symbol
- Gm11754
- Entrez Gene ID
- 105243636
- Gene Name
- predicted gene 11754
- Gene Aliases
- OTTMUSG00000004003
- Chromosome Location
- Chr. 11: 119105130 - 119111450 on Build GRCm38
- UniGene ID
- Mm.151218
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK149260.1 |
- |
1 |
230 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY742163.1 |
- |
1 |
230 |
|
Chr. 11 - 119106338 - 119111425 [-] |
GenBankmRNA |
BB642183.1 |
- |
1 |
230 |
|
Chr. 11 - 119106328 - 119111425 [-] |
View Full Product Details