Gene Transcripts
- Gene Symbol
- 1700003M07Rik
- Entrez Gene ID
- 72225
- Gene Name
- RIKEN cDNA 1700003M07 gene
- Gene Aliases
- Chromosome Location
- Chr. 4: 129960374 - 129965138 on Build GRCm38
- UniGene ID
- Mm.102193
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AI390380.1 |
- |
3 |
312 |
|
Chr. 4 - 129960850 - 129963657 [+] |
GenBankmRNA |
BY705842.1 |
- |
4 |
512 |
|
Chr. 4 - 129960376 - 129963520 [+] |
View Full Product Details