Gene Transcripts
- Gene Symbol
- 4930528J18Rik
- Entrez Gene ID
- 78195
- Gene Name
- RIKEN cDNA 4930528J18 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.243852
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019708.1 |
- |
1 |
43 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715488.1 |
- |
1 |
43 |
|
Chr. 10 - 117189846 - 117190267 [-] |
View Full Product Details