Gene Transcripts
- Gene Symbol
- Gm11426
- Entrez Gene ID
- 791422
- Gene Name
- predicted gene 11426
- Gene Aliases
- OTTMUSG00000000913
- Chromosome Location
- Chr. 11: 82633353 - 82636309 on Build GRCm38
- UniGene ID
- Mm.341562
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033582.1 |
|
3 |
1349 |
|
GenBank mRNA |
AY512914.1 |
- |
3 |
1349 |
|
AY512941.1 |
- |
3 |
1291 |
|
View Full Product Details