Gene Transcripts
- Gene Symbol
- 2310008N11Rik
- Entrez Gene ID
- 72316
- Gene Name
- RIKEN cDNA 2310008N11 gene
- Gene Aliases
- Chromosome Location
- Chr. 8: 26814595 - 26824423 on Build GRCm38
- UniGene ID
- Mm.490534
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045904.1 |
|
2 |
322 |
|
GenBank mRNA |
AK009242.1 |
- |
2 |
323 |
|
View Full Product Details