Gene Transcripts
- Gene Symbol
- MIR34AHG
- Entrez Gene ID
- 106614088
- Gene Name
- MIR34A host gene
- Gene Aliases
- Chromosome Location
- Chr. 1: 9148011 - 9182338 on Build GRCh38
- UniGene ID
- Hs.551837
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_132742.1 |
|
2 |
2503 |
|
GenBank mRNA |
EF570048.1 |
- |
2 |
2270 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000635687.1 |
- |
|
2557 |
|
Chr. - - [-] |
View Full Product Details