Gene Transcripts

Gene Symbol
LINC00442
Entrez Gene ID
348021
Gene Name
long intergenic non-protein coding RNA 442
Gene Aliases
Chromosome Location
Chr. 13: 19008259 - 19012634 on Build GRCh38
UniGene ID
Hs.314114
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026852.1 4 1714
GenBank mRNA BC047425.1 - 4 1714
BC067875.1 - 4 1714
BC094789.1 - 4 1714

View Full Product Details