Gene Transcripts
- Gene Symbol
- LINC01103
- Entrez Gene ID
- 101927360
- Gene Name
- long intergenic non-protein coding RNA 1103
- Gene Aliases
- Chromosome Location
- Chr. 2: 104488456 - 104510509 on Build GRCh38
- UniGene ID
- Hs.650324
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110113.1 |
|
6 |
945 |
|
GenBank mRNA |
BC040913.1 |
- |
6 |
945 |
|
View Full Product Details