Gene Transcripts

Gene Symbol
Lmln
Entrez Gene ID
239833
Gene Name
leishmanolysin-like (metallopeptidase M8 family)
Gene Aliases
5330415H22Rik, AI465495
Chromosome Location
Chr. 16: 33061871 - 33127695 on Build GRCm38
UniGene ID
Mm.131951
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172823.2 NP_766411.1 4, 5 419
GenBank mRNA AK030463.1 - 4, 5 420
BC111899.1 - 4, 5 420 40062192
BC112428.1 - 4, 5 420 40062196

View Full Product Details