Gene Transcripts
- Gene Symbol
- Defb10
- Entrez Gene ID
- 246085
- Gene Name
- defensin beta 10
- Gene Aliases
- BD-10, Defb7
- Chromosome Location
- Chr. 8: 21858901 - 21862011 on Build GRCm38
- UniGene ID
- Mm.220170
- Species
- Mus musculus
- Gene Symbol
- Defb11
- Entrez Gene ID
- 246081
- Gene Name
- defensin beta 11
- Gene Aliases
- BD-11, Defb8
- Chromosome Location
- Chr. 8: 21905374 - 21906432 on Build GRCm38
- UniGene ID
- Mm.221026
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK016897.1 |
- |
|
736 |
|
Chr. - - [-] |
View Full Product Details