Gene Transcripts

Gene Symbol
Olfr1404
Entrez Gene ID
258881
Gene Name
olfactory receptor 1404
Gene Aliases
MOR267-2
Chromosome Location
Chr. 1: 173215607 - 173216640 on Build GRCm38
UniGene ID
Mm.377733
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146881.2 NP_667092.1 1 532
GenBank mRNA BC120856.1 - 1 532

View Full Product Details