Gene Transcripts

Gene Symbol
Gm8369
Entrez Gene ID
666926
Gene Name
predicted gene 8369
Gene Aliases
EG666926
Chromosome Location
Chr. 19: 11491755 - 11512577 on Build GRCm38
UniGene ID
Mm.486382
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164202.1 NP_001157674.1 5 526

View Full Product Details