Gene Transcripts

Gene Symbol
Gm9376
Entrez Gene ID
668814
Gene Name
predicted gene 9376
Gene Aliases
EG668814
Chromosome Location
Chr. 14: 118267158 - 118267779 on Build GRCm38
UniGene ID
Mm.425909
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101609.1 NP_001095079.1 1 346

View Full Product Details