Gene Transcripts

Gene Symbol
Gabra3
Entrez Gene ID
24947
Gene Name
gamma-aminobutyric acid (GABA) A receptor, alpha 3
Gene Aliases
Chromosome Location
Chr. X: 152414332 - 152642650 on Build Rnor_6.0
UniGene ID
Rn.10367
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017069.3 NP_058765.3 8 1113
XM_006229511.2 XP_006229573.1 8 1225
GenBank mRNA CB725341.1 - 2 149
X51991.1 - 8 1120

View Full Product Details