Gene Transcripts

Gene Symbol
P2rx6
Entrez Gene ID
25041
Gene Name
purinergic receptor P2X, ligand-gated ion channel, 6
Gene Aliases
P2rxl1, P2x6, Rnap2x6
Chromosome Location
Chr. 11: 87434931 - 87445221 on Build Rnor_6.0
UniGene ID
Rn.10258
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012721.2 NP_036853.2 7 794
XM_008768851.1 XP_008767073.1 7 984
XM_008768852.1 XP_008767074.1 7 669
GenBank mRNA X92070.1 - 7 675
X97376.1 - 7 805

View Full Product Details