Gene Transcripts
- Gene Symbol
- Itgb3
- Entrez Gene ID
- 29302
- Gene Name
- integrin, beta 3
- Gene Aliases
- Chromosome Location
- Chr. 10: 92667869 - 92783413 on Build Rnor_6.0
- UniGene ID
- Rn.229225
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153720.1 |
NP_714942.1
|
8, 9 |
1126 |
|
GenBank mRNA |
AJ440952.1 |
- |
8, 9 |
1126 |
|
View Full Product Details