Gene Transcripts

Gene Symbol
Csrp2
Entrez Gene ID
29317
Gene Name
cysteine and glycine-rich protein 2
Gene Aliases
SmLIM
Chromosome Location
Chr. 7: 53630675 - 53649309 on Build Rnor_6.0
UniGene ID
Rn.94754
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177425.3 NP_803174.2 3 178
GenBank mRNA BC086356.1 - 3 192
CD373038.1 - 3 169
U44948.1 - 3 183

View Full Product Details