Gene Transcripts

Gene Symbol
Wisp2
Entrez Gene ID
29576
Gene Name
WNT1 inducible signaling pathway protein 2
Gene Aliases
CCN5
Chromosome Location
Chr. 3: 160207969 - 160219316 on Build Rnor_6.0
UniGene ID
Rn.53991
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031590.1 NP_113778.1 5 1262
GenBank mRNA AF259981.1 - 5 1262
BC089187.1 - 4 1072

View Full Product Details