Gene Transcripts

Gene Symbol
Stx8
Entrez Gene ID
59074
Gene Name
syntaxin 8
Gene Aliases
Chromosome Location
Chr. 10: 54512278 - 54753302 on Build Rnor_6.0
UniGene ID
Rn.20291
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031656.2 NP_113844.1 2 95
XM_008767863.1 XP_008766085.1 3 319
XM_008767864.1 XP_008766086.1 3 319
XM_008767865.1 XP_008766087.1 3 319
GenBank mRNA AF033109.1 - 2 100

View Full Product Details