Gene Transcripts

Gene Symbol
Cdx2
Entrez Gene ID
66019
Gene Name
caudal type homeo box 2
Gene Aliases
Chromosome Location
Chr. 12: 9464026 - 9470565 on Build Rnor_6.0
UniGene ID
Rn.64495
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023963.1 NP_076453.1 2, 3 798
XM_008768999.1 XP_008767221.1 2, 3 689
GenBank mRNA AJ278466.1 - 2, 3 798

View Full Product Details