Gene Transcripts

Gene Symbol
Kcnh4
Entrez Gene ID
114032
Gene Name
potassium channel, voltage gated eag related subfamily H, member 4
Gene Aliases
Bec2
Chromosome Location
Chr. 10: 88647564 - 88667382 on Build Rnor_6.0
UniGene ID
Rn.30035
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053630.1 NP_446082.1 2 378
GenBank mRNA AB022699.1 - 2 378
AJ007628.1 - 2 341

View Full Product Details